missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylcytosine (~90%), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Methylcytosine |
| CAS | 554-01-8 |
| Molecular Formula | C5H7N3O |
| Quantity | 5 g |
| Synonym | 2(1H)-Pyrimidinone, 6-amino-5-methyl-, 2(1H)-Pyrimidinone, 4-amino-5-methyl- (9CI), Cytosine, 5-methyl- (6CI,8CI), 6-Amino-5-methyl-2(1H)-pyrimidinone, 4-Amino-2-hydroxy-5-methylpyrimidine, 4-Amino-5-methyl-2(1H)-pyrimidinone, 5-Methylcytosine, NSC 137776 |
| InChI Formula | InChI=1S/C5H7N3O/c1-3-2-7-5(9)8-4(3)6/h2H,1H3,(H3,6,7,8,9) |
| SMILES | CC1=CNC(=O)N=C1N |
| IUPAC Name | 4-amino-5-methyl-1H-pyrimidin-2-one |
| Molecular Weight (g/mol) | 125.13 |
| Formula Weight | 125.0589 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction