missing translation for 'onlineSavingsMsg'
Learn More

5-Methylcytosine, MedChemExpress

Product Code. 16469326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469326 Supplier Medchem Express Supplier No. HYW008091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Methylcytosine is a well-characterized DNA modification, and is also predominantly in abundant non-coding RNAs in both prokaryotes and eukaryotes. 5-Methylcytosine in mRNA is a new epitranscriptome marker inArabidopsis, and that regulation of this modification is an integral part of gene regulatory networks underlying plant development.

5-Methylcytosine is a well-characterized DNA modification, and is also predominantly in abundant non-coding RNAs in both prokaryotes and eukaryotes. 5-Methylcytosine in mRNA is a new epitranscriptome marker inArabidopsis, and that regulation of this modification is an integral part of gene regulatory networks underlying plant development.

Specifications

Chemical Name or Material 5-Methylcytosine
CAS 554-01-8
Color White
Purity Grade Notes Research
Molecular Formula C5H7N3O
Quantity 100 mg
Solubility Information DMSO : 8.33 mg/mL (66.57 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (57.06 mM; Need ultrasonic)
SMILES O=C1N=CC(C)=C(N)N1
Molecular Weight (g/mol) 125.13
Formula Weight 125.13
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.