missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfuran-2-boronic acid pinacol ester, 97%

Product Code. 15474057
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474057 1 g 1g
15484057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474057 Supplier Thermo Scientific Alfa Aesar Supplier No. H31282.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 338998-93-9 | C11H17BO3 | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338998-93-9
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.06
MDL Number MFCD03094689
InChI Key FNPZFZKLYGWKLH-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-5-methylfuran-2-yl-1,3,2-dioxaborolane, 5-methylfuran-2-boronic acid pinacol ester, 2-methylfurane-5-boronic acid pinacol ester, 2-methylfuran-5-boronic acid pinacol ester, 2-methyl-furyl-5-pinacolato boron, 5-methyl-2-furanboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-methyl-2-furyl-1,3,2-dioxaborolane, 5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxoborolan-2-yl furan, 4,4,5,5-tetramethyl-2-5-methyl-2-furanyl-1,3,2-dioxaborolane, 5-methylfuran-2-yl boronic acid pinacol ester
PubChem CID 2736881
IUPAC Name 4,4,5,5-tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane
SMILES CC1=CC=C(O1)B1OC(C)(C)C(C)(C)O1

Specifications

Density 1.023
Quantity 1 g
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 5-Methylfuran-2-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.