missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, TRC

Product Code. 30012847
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012847 5 g 5g
30003483 100 mg 100mg
30006016 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012847 Supplier Toronto Research Chemicals Supplier No. TRCM3054705G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6 H6 O2
Molecular Weight (g/mol) 110.11
Synonym 5-Methyl-2-furfural, 5-Methylfuran-2-carbaldehyde, 2-Furaldehyde, 5-methyl- (6CI,8CI), 5-Methyl-2-furancarboxaldehyde, 2-Formyl-5-methylfuran, 2-Formyl-5-methyltetrahydrofuran, 2-Methyl-5-formylfuran, 2-Methyl-5-furaldehyde, 5-Methyl-2-furaldehyde, 5-Methyl-2-furfural, 5-Methyl-2-furfuraldehyde, 5-Methyl-2-furfurylaldehyde, 5-Methylfuran-2-al, 5-Methylfuran-2-aldehyde, 5-Methylfurfural, 5-Methylfurfuraldehyde
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES Cc1oc(C=O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H6O2/c1-5-2-3-6(4-7)8-5/h2-4H,1H3
Formula Weight 110.0368
Percent Purity >95%
Chemical Name or Material 5-Methyl-2-furfural
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.