missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisatin, 95%

Product Code. 15364978 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15364978 5 g 5g
15354978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15364978 Supplier Thermo Scientific Acros Supplier No. 206860050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 608-05-9 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-05-9
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005721
InChI Key VAJCSPZKMVQIAP-UHFFFAOYSA-N
Synonym 5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
PubChem CID 11840
IUPAC Name 5-methyl-1H-indole-2,3-dione
SMILES CC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Beilstein 21,509
Solubility Information Solubility in water: insoluble.
Formula Weight 161.16
Percent Purity 95%
Chemical Name or Material 5-Methylisatin
compliance-icons
Product Identifier
  • 5-Methylisatin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.