missing translation for 'onlineSavingsMsg'
Learn More

(5-Methylisoxazol-3-yl)methanol, 97%, Thermo Scientific™

Product Code. 10222732
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10222732 1 g 1g
10167384 5 g 5g
10499644 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10222732 Supplier Thermo Scientific Maybridge Supplier No. CC10409DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35166-33-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00085129
InChI Key MDYHWQQHEWDJKR-UHFFFAOYSA-N
Synonym 5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
PubChem CID 2736567
IUPAC Name (5-methyl-1,2-oxazol-3-yl)methanol
SMILES CC1=CC(CO)=NO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 113.12
Percent Purity 97%
Chemical Name or Material (5-Methylisoxazol-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.