missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyrazin-2-ylamine, 97%, Thermo Scientific™

Product Code. 10672295
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10672295 5 g Amber Glass Bottle 5g
10547284 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10672295 Supplier Thermo Scientific Maybridge Supplier No. MO08458DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5521-58-4
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD08437664
InChI Key ZNQOALAKPLGUPH-UHFFFAOYSA-N
Synonym 2-amino-5-methylpyrazine, 5-methyl-pyrazin-2-ylamine, 5-methyl-2-pyrazinamine, pyrazinamine, 5-methyl, 2-amino-5 methylpyrazine, 5-methyl-2-aminopyrazine, 2-amino-5-methyl pyrazine, 5-methylpyrazine-2-ylamine, 2-pyrazinamine, 5-methyl, pubchem15710
PubChem CID 313215
IUPAC Name 5-methylpyrazin-2-amine
SMILES CC1=CN=C(C=N1)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 5-Methylpyrazin-2-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.