missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylsulfonyl)pyridine-3-boronic acid, 98%

Product Code. 15410416
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410416 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410416 Supplier Thermo Scientific Alfa Aesar Supplier No. H53133.MD
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 913836-01-8 | C6H8BNO4S | 201.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 913836-01-8
Melting Point 123°C to 125°C
Molecular Formula C6H8BNO4S
MDL Number MFCD08689536
Quantity 250 mg
Synonym 5-methylsulphonyl pyridine-3-boronic acid, 5-methylsulfonyl pyridine-3-boronic acid, 5-methylsulfonyl pyridin-3-yl boronic acid, 5-methylsulfonyl-3-pyridineboronic acid, boronicacid, b-5-methylsulfonyl-3-pyridinyl, 5-methanesulfonylpyridin-3-ylboronic acid, b-5-methylsulfonyl-3-pyridinyl boronic acid, acmc-209ram
InChI Key YGXNPQDXWDCWET-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CN=CC(=C1)B(O)O
IUPAC Name (5-methanesulfonylpyridin-3-yl)boronic acid
Molecular Weight (g/mol) 201.00
PubChem CID 44119381
Formula Weight 201
Percent Purity 98%
Chemical Name or Material 5-(Methylsulfonyl)pyridine-3-boronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.