missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylsulfonyl)thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10757685
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10757685 10 g Amber Glass Bottle 10g
10111282 250 mg Amber Glass Bottle 250mg
10120562 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10757685 Supplier Thermo Scientific Maybridge Supplier No. SEW03813EA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 60166-86-1
Molecular Formula C6H6O4S2
Molecular Weight (g/mol) 206.23
MDL Number MFCD00173745
InChI Key SLWRTVINHWIGTK-UHFFFAOYSA-N
Synonym 5-methylsulfonyl thiophene-2-carboxylic acid, 5-methanesulfonylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-methylsulfonyl, maybridge1_007675, 5-methanesulfonyl-2-thiphenecarboxylic acid, 5-methylsulfonyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid,5-methylsulfonyl, 5-methanesulfonyl thiophene-2-carboxylic acid
PubChem CID 2740497
IUPAC Name 5-methylsulfonylthiophene-2-carboxylic acid
SMILES CS(=O)(=O)C1=CC=C(S1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 5-(Methylsulfonyl)thiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.