missing translation for 'onlineSavingsMsg'
Learn More

(5-Methylthien-2-yl)methylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10764467
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10764467 5 g Amber Glass Bottle 5g
10643265 250 mg Amber Glass Bottle 250mg
10268003 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10764467 Supplier Thermo Scientific Maybridge Supplier No. CC65063DE
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 171661-55-5
Molecular Formula C6H10ClNS
Molecular Weight (g/mol) 163.663
InChI Key COTMVCYERKXKDX-UHFFFAOYSA-N
Synonym 5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthien-2-yl methylamine hydrochloride, 5-methylthien-2-yl methylamine hcl, 1-5-methylthiophen-2-yl methanamine hydrochloride, 5-methylthiophen-2-yl methanaminehydrochloride, 5-methylthien-2-yl methylamine-hcl, 5-methyl-2-thienyl methylamine, chloride, 2-aminomethyl-5-methylthiophene hydrochloride, 2-thiophenemethanamine, 5-methyl-, hydrochloride, 2-thiophenemethanamine,5-methyl-,hydrochloride 1:1
PubChem CID 24229725
IUPAC Name (5-methylthiophen-2-yl)methanamine;hydrochloride
SMILES CC1=CC=C(S1)CN.Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 163.67
Percent Purity 97%
Chemical Name or Material (5-methylthien-2-yl)methylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.