missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10747305
Change view
Click to view available options
Quantity:
250 mg
1 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10747305 50 g Amber Glass Bottle 50g
10088291 250 mg Amber Glass Bottle 250mg
10213572 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10747305 Supplier Thermo Scientific Maybridge Supplier No. SEW03638EE
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 20873-58-9
Molecular Formula C6H6O2S2
Molecular Weight (g/mol) 174.232
MDL Number MFCD00173742
InChI Key FWFWUSLBIIIIEN-UHFFFAOYSA-N
Synonym 5-methylthio thiophene-2-carboxylic acid, 5-methylsulfanyl thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-methylthio, 2-thiophenecarboxylicacid, 5-methylthio, 5-methylthiothiophene-2-carboxylic acid, maybridge1_007717, d04ngc, cambridge id 5103856, 5-methylmercaptothiophene-2-carboxylic, 5-methylthio-2-thiophenecarboxylic acid
PubChem CID 714372
IUPAC Name 5-methylsulfanylthiophene-2-carboxylic acid
SMILES CSC1=CC=C(S1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 174.24
Percent Purity 97%
Chemical Name or Material 5-(Methylthio)thiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.