missing translation for 'onlineSavingsMsg'
Learn More

5'-Methylthioadenosine, MedChemExpress

Product Code. 16453556
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453556 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453556 Supplier Medchem Express Supplier No. HY16938
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5'-Methylthioadenosine (5'-(Methylthio)-5'-deoxyadenosine) is a nucleoside generated from S-adenosylmethionine (SAM) during polyamine synthesis.

5'-Methylthioadenosine suppresses tumors by inhibiting tumor cell proliferation, invasion, and the induction of apoptosis while controlling the inflammatory micro-environments of tumor tissue. 5'-Methylthioadenosine and its associated materials have striking regulatory effects on tumorigenesis.

Specifications

Chemical Name or Material 5'-Methylthioadenosine
CAS 2457-80-9
Color White
Purity Grade Notes Research
Molecular Formula C11H15N5O3S
Synonym 5'-(Methylthio)-5'-deoxyadenosine 5'-Deoxy-5'-(methylthio)adenosine 5'-S-Methyl-5'-thioadenosine
Solubility Information DMSO : 50 mg/mL (168.16 mM; Need ultrasonic) ∣DMF : 2.5 mg/mL (8.41 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.36 mM; Need ultrasonic)
SMILES O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3CSC
Molecular Weight (g/mol) 297.33
Quantity 25 mg
Formula Weight 297.33
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.