missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10584994
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10584994 250 mg Amber Glass Bottle 250mg
10358353 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10584994 Supplier Thermo Scientific Maybridge Supplier No. CC65002CB
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 31555-59-6
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.615
MDL Number MFCD00130092
InChI Key DJCGIWVUKCNCAT-UHFFFAOYSA-N
Synonym 5-methyl-thiophene-2-carbonyl chloride, 5-methyl-2-thiophenecarbonyl chloride, 2-thiophenecarbonyl chloride, 5-methyl, 2-thiophenecarbonylchloride, 5-methyl, 5-methyl-2-thenoyl chloride, 2-chlorocarbonyl-5-methylthiophene, 5-methyl-thiophene-carboxylicacid chloride, 5-methylthiophene-2-carboxylic acid chloride, 5-methyl-thiophene-2-carboxylic acid chloride
PubChem CID 2760083
IUPAC Name 5-methylthiophene-2-carbonyl chloride
SMILES CC1=CC=C(S1)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 160.62
Percent Purity 97%
Chemical Name or Material 5-Methylthiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.