missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylthiophene-2-carboxaldehyde, 98+%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.
Solubility
Soluble in ether and ethanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.
Solubility
Soluble in ether and ethanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.18 |
| Boiling Point | 92°C to 93°C (12 mmHg) |
| Flash Point | 87°C (188°F) |
| Refractive Index | 1.583 |
| Quantity | 10 g |
| Beilstein | 106896 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in ether and ethanol. |
| Formula Weight | 126.18 |
| Percent Purity | ≥98% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction