missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10387304
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10387304 1 g 1g
10673265 5 g 5g
10596334 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10387304 missing translation for 'mfr' Thermo Scientific Maybridge missing translation for 'supplierNo' CC65003DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 55854-45-0
Molecular Formula C5H5ClO2S2
Molecular Weight (g/mol) 196.66
MDL Number MFCD05864499
InChI Key QGGRRHYGHGJEKP-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
PubChem CID 12400112
IUPAC Name 5-methylthiophene-2-sulfonyl chloride
SMILES CC1=CC=C(S1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 196.68
Percent Purity 97%
Chemical Name or Material 5-Methylthiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.