missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KYP 2047

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
KYP 2047 is a high affinity prolyl oligopeptidase (POP) inhibitor (Ki = 0.023 nM). Clears α-synuclein aggregates induced by oxidative stress in neuronal cells. Inhibits the formation of AcSDKP from its precursor 43-mer thymosin β4 (Tβ4). Induces angiogenesis via POP inhibition. It reduces tau aggregation in tau-transfected HEK-293 cells and N2A cells as well as in human iPSC-derived neurons from patients with frontotemporal dementia. In vivo, treatment with KYP 2047 of transgenic mice with tauopathy, reduces tau aggregation in the brain and cerebrospinal fluid and slows cognitive decline. Anti-angiogenic and neuroprotective. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | (2S)-1-[[(2S)-1-(1-Oxo-4-phenylbutyl)-2-pyrrolidinyl]carbonyl]-2-pyrrolidinecarbonitrile |
| CAS | 796874-99-2 |
| Quantity | 5 mg |
| Molecular Formula | C20H25N3O2 |
| Formula Weight | 339.44 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction