missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)-2-furaldehyde, 97%, Thermo Scientific™

Product Code. 10515934
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10515934 250 mg Amber Glass Bottle 250mg
10396654 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515934 Supplier Thermo Scientific Maybridge Supplier No. CC72504CB
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 392659-97-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD01821235
InChI Key ZQILSGYYJOBENS-UHFFFAOYSA-N
Synonym 5-morpholinomethyl-2-furaldehyde, 5-morpholin-4-ylmethyl furan-2-carbaldehyde, 5-morpholin-4-ylmethyl-furan-2-carbaldehyde, 5-morpholin-4-ylmethyl furan-2-carboxaldehyde, 2-furancarboxaldehyde,5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl-2-furaldehyde, 5-morpholinomethyl furan-2-carbaldehyde, 5-morpholin-4-yl methyl furan-2-carbaldehyde
PubChem CID 3159700
IUPAC Name 5-(morpholin-4-ylmethyl)furan-2-carbaldehyde
SMILES C1COCCN1CC2=CC=C(O2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 195.22
Percent Purity 97%
Chemical Name or Material 5-(Morpholinomethyl)-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.