missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1,10-phenanthroline, 98%

Product Code. 11480273
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480273 25 g 25g
11460273 1 g 1g
11470273 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480273 Supplier Thermo Scientific Alfa Aesar Supplier No. A11428.14
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4199-88-6 | C12H7N3O2 | 225.207 g/mol

5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4199-88-6
Molecular Formula C12H7N3O2
Molecular Weight (g/mol) 225.207
MDL Number MFCD00004981
InChI Key PDDBTWXLNJNICS-UHFFFAOYSA-N
PubChem CID 72790
IUPAC Name 5-nitro-1,10-phenanthroline
SMILES C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]

Specifications

Melting Point 202°C to 204°C
Quantity 25 g
Beilstein 196245
Synonym 1,10-phenanthroline, 5-nitro, 5-nitro-1, 10-diazaphenanthrene, nitroferroin, 5-nitropyridino 3,2-h quinoline, acmc-209jmn, phhn01, 5nitro-1, 10-phenanthroline, 5-nitro-1,10-diazaphenanthrene, pddbtwxlnjnics-uhfffaoysa, 5-nitro-1,10-phenanthroline, crystalline
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 225.21
Percent Purity 98%
Chemical Name or Material 5-Nitro-1,10-phenanthroline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.