missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1H-indole, 97%, Thermo Scientific™

Product Code. 10482124
Change view
Click to view available options
:
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10482124 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482124 Supplier Thermo Scientific Maybridge Supplier No. SB01605EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6146-52-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6N2O2
MDL Number MFCD00005673
Quantity 10g
Synonym 5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr
InChI Key OZFPSOBLQZPIAV-UHFFFAOYSA-N
SMILES C1=CC2=C(C=CN2)C=C1[N+](=O)[O-]
IUPAC Name 5-nitro-1H-indole
Molecular Weight (g/mol) 162.148
PubChem CID 22523
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 5-Nitro-1H-indole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.