missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-benzimidazolinone, 99%

Product Code. 11369145
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369145 5 g 5g
11359145 1 g 1g
11379145 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369145 Supplier Thermo Scientific Alfa Aesar Supplier No. L16391.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-84-5 | C7H5N3O3 | 179.14 g/mol

5-Nitro-2-benzimidazolinone is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-benzimidazolinone is used as intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-84-5
Molecular Formula C7H5N3O3
Molecular Weight (g/mol) 179.14
MDL Number MFCD00220274
InChI Key DLJZIPVEVJOKHB-UHFFFAOYSA-N
Synonym 5-nitro-2-benzimidazolinone, 5-nitro-1h-benzo d imidazol-2 3h-one, 5-nitrobenzimidazol-2-one, 5-nitro-1,3-dihydro-2h-benzimidazol-2-one, 2-hydroxy-5-nitrobenzimidazole, 5-nitro-2 3h-benzimidazolone, 2h-benzimidazol-2-one, 1,3-dihydro-5-nitro, 2-benzimidazolinone, 5-nitro, 5-nitro-1h-benzimidazol-2-ol, 1,3-dihydro-5-nitro-2h-benzimidazol-2-one
PubChem CID 3725611
SMILES [O-][N+](=O)C1=CC=C2NC(=O)NC2=C1

Specifications

Melting Point >300°C
Quantity 5 g
UN Number UN2811
Beilstein 171386
Solubility Information Insoluble in water.
IUPAC Name 5-nitro-1,3-dihydrobenzimidazol-2-one
Formula Weight 179.14
Percent Purity 99%
Chemical Name or Material 5-Nitro-2-benzimidazolinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.