Learn More
5-Nitro-2-furaldehyde semicarbazone, 98+%, Thermo Scientific™
27.70 € - 131.00 €
Chemical Identifiers
| CAS | 59-87-0 |
|---|---|
| Molecular Formula | C6H6N4O4 |
| Molecular Weight (g/mol) | 198.14 |
| MDL Number | MFCD00003225 |
| InChI Key | IAIWVQXQOWNYOU-FPYGCLRLSA-N |
| Synonym | nitrofurazone, nitrofural, furacilin, furacin, furacillin, aldomycin, furacine, furaldon, nifuzon, actin-n |
| PubChem CID | 5447130 |
| IUPAC Name | [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea |
| SMILES | NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11364649
|
Thermo Scientific Alfa Aesar
A18593.14 |
25g |
27.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11374649
|
Thermo Scientific Alfa Aesar
A18593.22 |
100g |
65.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11428517
|
Thermo Scientific Alfa Aesar
A18593.30 |
250g |
131.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H6N4O4 | |
| MFCD00003225 | |
| nitrofurazone, nitrofural, furacilin, furacin, furacillin, aldomycin, furacine, furaldon, nifuzon, actin-n | |
| [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea |
Specifications
| 59-87-0 | |
| MFCD00003225 | |
| Light sensitive | |
| nitrofurazone, nitrofural, furacilin, furacin, furacillin, aldomycin, furacine, furaldon, nifuzon, actin-n | |
| NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O | |
| 198.14 | |
| 198.14 | |
| 5-Nitro-2-furaldehyde semicarbazone |
| C6H6N4O4 | |
| 86403 | |
| 14,6600 | |
| IAIWVQXQOWNYOU-FPYGCLRLSA-N | |
| [(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea | |
| 5447130 | |
| ≥98% |
Safety and Handling
GHS H Statement
H301-H341-H351-H361
Toxic if swallowed.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
missing translation for 'einecsNumber' : 200-443-1
missing translation for 'rtecsNumber' : LT7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.