missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-indanone, 97%, Thermo Scientific™

Product Code. 10225564
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10225564 5 g Amber Glass Bottle 5g
10215564 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10225564

Brand: Thermo Scientific Maybridge MO08378DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 116530-60-0
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00598938
InChI Key VSEBFWRYDORZJI-UHFFFAOYSA-N
PubChem CID 1502066
IUPAC Name 5-nitro-1,3-dihydroinden-2-one
SMILES [O-][N+](=O)C1=CC2=C(CC(=O)C2)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 5-nitro-2-indanone, 5-nitro-1h-inden-2 3h-one, 5-nitro-indan-2-one, 5-nitroindan-2-one, 2h-inden-2-one, 1,3-dihydro-5-nitro, 5-nitro-1,3-dihydro-2h-inden-2-one, pubchem18090, acmc-2099su, 2h-inden-2-one,1,3-dihydro-5-nitro, 1,3-dihydro-5-nitro-2h-inden-2-one
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.