missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-7-azaindole, 97%, Thermo Scientific Chemicals

Product Code. 15425947
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425947 250 mg 250mg
15415947 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425947 Supplier Thermo Scientific Alfa Aesar Supplier No. H33141.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101083-92-5
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD06659682
InChI Key INMIPMLIYKQQID-UHFFFAOYSA-N
Synonym 5-nitro-7-azaindole, 5-nitro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 5-nitro, pubchem17751, acmc-1bu2p, 1h-pyrrolo 2,3-b pyridine,5-nitro
PubChem CID 11521140
IUPAC Name 5-nitro-1H-pyrrolo[2,3-b]pyridine
SMILES [O-][N+](=O)C1=CN=C2NC=CC2=C1

Specifications

Melting Point 280°C
Quantity 250 mg
Formula Weight 163.14
Percent Purity 97%
Chemical Name or Material 5-Nitro-7-azaindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.