missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzimidazole, 98+%

Product Code. 11448793
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448793 50 g 50g
11458793 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448793 Supplier Thermo Scientific Alfa Aesar Supplier No. A10524.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-52-0 | C7H5N3O2 | 163.14 g/mol

5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.

Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-52-0
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00005604
InChI Key XPAZGLFMMUODDK-UHFFFAOYSA-N
Synonym 5-nitrobenzimidazole, 6-nitrobenzimidazole, 5-nitro-1h-benzo d imidazole, 5-nitro-1h-benzimidazole, 1h-benzimidazole, 5-nitro, 1h-benzimidazole, 6-nitro, 6-nitro-benzimidazole, 5 6-nitrobenzimidazole, benzimidazole, 5-nitro, benzimidazole, 6-nitro
PubChem CID 7195
IUPAC Name 6-nitro-1H-benzimidazole
SMILES [O-][N+](=O)C1=CC=C2N=CNC2=C1

Specifications

Melting Point 207°C to 211°C
Quantity 50 g
Beilstein 7926
Solubility Information Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.
Formula Weight 163.14
Percent Purity ≥98%
Chemical Name or Material 5-Nitrobenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.