missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzothiazole, 96%, Thermo Scientific Chemicals

Product Code. 15456437
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456437 1 g 1g
15466437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456437 Supplier Thermo Scientific Alfa Aesar Supplier No. H33594.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2942-07-6 | Molecular Formula: C12H18ClNO2 | Molecular Weight: 243.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-07-6
MDL Number MFCD00085891

Specifications

Melting Point 157°C to 162°C
Molecular Formula C12H18ClNO2
Quantity 1 g
Synonym benzoylcholine chloride, 2-hydroxyethyl trimethylammonium chloride benzoate, choline, chloride, benzoate, 2-benzoyloxy-n,n,n-trimethylethanaminium chloride, 2-benzoyloxyethyltrimethylammonium chloride, 2-benzoyloxy ethyl trimethylazanium chloride, 2-benzyloxy-n,n,n-trimethylethanaminium chloride, 2-benzyloxy-n,n,n-trimethylethanaminimum chloride, ammonium, 2-hydroxyethyl trimethyl-, chloride, benzoate, 2-trimethylamino ethyl benzoate, chloride
InChI Key QVFHQENRNSAHEK-UHFFFAOYSA-M
SMILES C[N+](C)(C)CCOC(=O)C1=CC=CC=C1.[Cl-]
IUPAC Name 2-benzoyloxyethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 243.73
PubChem CID 76298
Formula Weight 180.18
Percent Purity 96%
Chemical Name or Material 5-Nitrobenzothiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.