missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrofuraldehyde Diacetate, TRC

Product Code. 30137612
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137612 1 g 1g
30136519 500 mg 500mg
30011640 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137612 Supplier Toronto Research Chemicals Supplier No. TRCN4959001G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-55-7
Molecular Formula C9 H9 N O7
Molecular Weight (g/mol) 243.17
Synonym (5-Nitrofuran-2-yl)methylidene Diacetate, Nitrofural Imp. B (EP), Nifuroxazide Imp. C (EP)
IUPAC Name [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
SMILES CC(=O)OC(OC(=O)C)c1oc(cc1)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H9NO7/c1-5(11)15-9(16-6(2)12)7-3-4-8(17-7)10(13)14/h3-4,9H,1-2H3
Formula Weight 243.0379
Chemical Name or Material (5-Nitrofuran-2-yl)methylene Diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.