missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carbonitrile, 98+%

Product Code. 11307526
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307526 1 g 1g
11317526 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307526 Supplier Thermo Scientific Alfa Aesar Supplier No. L11058.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 16689-02-4 | C5H2N2O2S | 154.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16689-02-4
Molecular Formula C5H2N2O2S
Molecular Weight (g/mol) 154.143
MDL Number MFCD00052401
InChI Key FLYONFCGDKAMIH-UHFFFAOYSA-N
Synonym 5-nitro-2-thiophenecarbonitrile, 5-cyano-2-nitrothiophene, 2-cyano-5-nitrothiophene, 5-nitrothiophene-2-carbonitirile, 5-nitro-thiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-nitro, zlchem 478, maybridge1_005980, 5-nitro-2-cyanothiophene, acmc-209dv8
PubChem CID 519294
IUPAC Name 5-nitrothiophene-2-carbonitrile
SMILES C1=C(SC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 43°C to 45°C
Quantity 1 g
UN Number UN3439
Beilstein 123934
Formula Weight 154.15
Percent Purity ≥98%
Chemical Name or Material 5-Nitrothiophene-2-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.