missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carboxaldehyde, 98%

Product Code. 11371297
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371297 5 g 5g
11381297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371297 Supplier Thermo Scientific Alfa Aesar Supplier No. L01493.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4521-33-9 | C5H3NO3S | 157.143 g/mol

5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4521-33-9
Molecular Formula C5H3NO3S
Molecular Weight (g/mol) 157.143
MDL Number MFCD00005433
InChI Key CHTSWZNXEKOLPM-UHFFFAOYSA-N
Synonym 5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene
PubChem CID 78281
IUPAC Name 5-nitrothiophene-2-carbaldehyde
SMILES C1=C(SC(=C1)[N+](=O)[O-])C=O

Specifications

Melting Point 72°C to 76°C
Quantity 5 g
Beilstein 120540
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 157.15
Percent Purity 98%
Chemical Name or Material 5-Nitrothiophene-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.