missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carboxylic acid, 98%

Product Code. 15472199
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15472199 100 g 100g
15452199 5 g 5g
15462199 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15472199 Supplier Thermo Scientific Alfa Aesar Supplier No. H64821.22
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6317-37-9 | C5H3NO4S | 173.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6317-37-9
Molecular Formula C5H3NO4S
Molecular Weight (g/mol) 173.142
MDL Number MFCD00159552
InChI Key UNEPVPOHGXLUIR-UHFFFAOYSA-N
Synonym 5-nitrothiophene-2-carboxylic acid, 5-nitro-2-thiophenecarboxylic acid, 5-nitro-2-thenoic acid, 5-nitrothiophene-2-carboxylicacid, 5-nitro-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-nitro, 2-carboxy-5-nitrothiophene, pubchem5568, acmc-1b47f, 5-nitrothiophene-2-carboxylic
PubChem CID 80591
SMILES C1=C(SC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 154°C to 159°C
Color Yellow
Quantity 100 g
Beilstein 139665
Formula Weight 173.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Nitrothiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.