missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanol, 97%, Thermo Scientific™

Product Code. 15215998 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 15215998

Brand: Thermo Scientific Acros 390000050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 623-93-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
InChI Key FCBBRODPXVPZAH-UHFFFAOYSA-N
Synonym 5-nonanol, dibutylcarbinol, dibutyl carbinol, 5-nonyl alcohol, nonanol-5, acmc-1bc2t, di-n-butyl carbinol, 5-nonanol 25g, 5-nonanol gc, 5-nonanol, puriss gc
PubChem CID 12202
IUPAC Name nonan-5-ol
SMILES CCCCC(CCCC)O

Specifications

Melting Point 5.0°C
Density 0.8200g/mL
Boiling Point 194.0°C to 196.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
Refractive Index 1.429
Quantity 5 mL
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material 5-Nonanol
Show More Show Less
compliance-icons
Product Identifier
  • 5-Nonanol, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.