missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone, 98%

Product Code. 10457000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Förpackningsstorlek:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
10457000 25 mL Glass bottle 25mL
10551601 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10457000 Leverantör Thermo Scientific Acros Leverantörsnummer 159760250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 502-56-7 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifikationer

CAS Min % 97.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 186.0°C to 187.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CO(CH2)3CH3
Refractive Index 1.4180 to 1.4200
Quantity 25 mL
Beilstein 01, I, 365
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol;, very soluble in diethyl ether, chloroform.
Formula Weight 142.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Nonanone, 98%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.