missing translation for 'onlineSavingsMsg'
Learn More

5'-O-Acetyl Adenosine, TRC

Product Code. 30309876
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309876 Supplier Toronto Research Chemicals Supplier No. TRCA168060500MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5'-O-Acetyl Adenosine
CAS 2140-25-2
Molecular Formula C12 H15 N5 O5
Quantity 500 mg
Synonym Adenosine 5'-Acetate, 5'-Acetyladenosine, NSC 91775
InChI Formula InChI=1S/C12H15N5O5/c1-5(18)21-2-6-8(19)9(20)12(22-6)17-4-16-7-10(13)14-3-15-11(7)17/h3-4,6,8-9,12,19-20H,2H2,1H3,(H2,13,14,15)/t6-,8-,9-,12-/m1/s1
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl acetate
Molecular Weight (g/mol) 309.28
Formula Weight 309.1073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.