missing translation for 'onlineSavingsMsg'
Learn More

5-(o-Tolyl)-1,3,4-oxadiazole-2-thiol, 96%, Thermo Scientific™

Product Code. 15496627
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496627 1 g 1g
15406637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496627 Supplier Thermo Scientific Alfa Aesar Supplier No. H33776.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2503-66-4
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.236
MDL Number MFCD00992514
InChI Key HNFIJWNVPWXSOL-UHFFFAOYSA-N
Synonym 5-2-methylphenyl-1,3,4-oxadiazole-2-thiol, 5-o-tolyl-3h-1,3,4 oxadiazole-2-thione, 5-o-tolyl-1,3,4-oxadiazole-2-thiol, 5-2-methylphenyl-3h-1,3,4-oxadiazole-2-thione, 1,3,4-oxadiazole-2 3h-thione,5-2-methylphenyl, 5-o-tolyl-3h-1,3,4-oxadiazole-2-thione, 5-2-methylphenyl-1,3,4-oxadiazoline-2-thione, 5-2-methylphenyl-1,3,4-oxadiazol-2 3h-thione
PubChem CID 774385
IUPAC Name 5-(2-methylphenyl)-3H-1,3,4-oxadiazole-2-thione
SMILES CC1=CC=CC=C1C2=NNC(=S)O2

Specifications

Melting Point 181°C to 182°C
Quantity 1 g
Formula Weight 192.24
Percent Purity 96%
Chemical Name or Material 5-(o-Tolyl)-1,3,4-oxadiazole-2-thiol
Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.