missing translation for 'onlineSavingsMsg'
Learn More

5-[p-(Dimethylamino)benzylidene]rhodanine, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15528784 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15528784 10 g Wide Mouth Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528784 Supplier Avantor J.T.Baker Supplier No. J43102

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 536-17-4
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C12H12N2OS2
Quantity 10 g
Synonym unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
InChI Key JJRVRELEASDUMY-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol) 264.361
PubChem CID 2723826
Formula Weight 264.37
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material 5-[p-(Dimethylamino)benzylidene]rhodanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.