missing translation for 'onlineSavingsMsg'
Learn More

5-(p-Methylphenyl)-5-phenylhydantoin, TRC

Product Code. 30134455
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134455 1 g 1g
30012828 100 mg 100mg
30010617 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134455 Supplier Toronto Research Chemicals Supplier No. TRCM3206151G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51169-17-6
Molecular Formula C16 H14 N2 O2
Molecular Weight (g/mol) 266.29
Synonym 5-(4-Methylphenyl)-5-phenylimidazolidine-2,4-dione, Phenytoin Imp. F (EP), Phenytoin Sodium Imp. F (EP)
IUPAC Name 5-(4-methylphenyl)-5-phenylimidazolidine-2,4-dione
SMILES Cc1ccc(cc1)C2(NC(=O)NC2=O)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H14N2O2/c1-11-7-9-13(10-8-11)16(12-5-3-2-4-6-12)14(19)17-15(20)18-16/h2-10H,1H3,(H2,17,18,19,20)
Formula Weight 266.1055
Percent Purity >95%
Chemical Name or Material 5-(4-Methylphenyl)-5-phenylimidazolidine-2,4-dione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.