missing translation for 'onlineSavingsMsg'
Learn More

5-Phenoxy-2-furoic acid, ≥97%, Thermo Scientific™

Product Code. 10659325
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10659325 5 g 5g
10600665 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10659325 Supplier Thermo Scientific Maybridge Supplier No. HTS06019DE
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 60698-32-0
Molecular Formula C11H8O4
Molecular Weight (g/mol) 204.18
MDL Number MFCD01925067
InChI Key QQKXTFLRAZKJJO-UHFFFAOYSA-N
Synonym 5-phenoxy-2-furoic acid, 2-furancarboxylic acid,5-phenoxy, 5-phenoxy-furan-2-carboxylic acid, maybridge3_002285, 5-phenoxypyromucic acid
PubChem CID 2812790
IUPAC Name 5-phenoxyfuran-2-carboxylic acid
SMILES OC(=O)C1=CC=C(OC2=CC=CC=C2)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 204.18
Percent Purity 97%
Chemical Name or Material 5-Phenoxy-2-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.