missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1H-tetrazole, TRC

Product Code. 30428375
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428375 250 mg -
30422695 500 mg -
30418268 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428375 Supplier Toronto Research Chemicals Supplier No. TRCP337023250MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18039-42-4
Molecular Formula C7 H6 N4
Molecular Weight (g/mol) 146.15
Synonym 5-Phenyl-2H-tetrazole, 1H-Tetrazole, 5-phenyl- (8CI,9CI), Tetrazole, 5-phenyl- (6CI), 5-Phenyl-1,2,3,4-tetrazole, 5-Phenyl-1H-tetrazole, 5-Phenyl-2H-tetrazole, 5-Phenyltetrazole, Celltetra BHT-P 5T, Celltetra P 5T, Expandex 5PT, Expandex OX 5PT, Kempore 50XPT, MA 1623, NSC 11138, Phenyltetrazole, Unicell 5PT, Unifoam HB 1
IUPAC Name 5-phenyl-1H-tetrazole
SMILES c1ccc(cc1)c2nnn[nH]2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H6N4/c1-2-4-6(5-3-1)7-8-10-11-9-7/h1-5H,(H,8,9,10,11)
Formula Weight 146.0592
Chemical Name or Material 5-Phenyl-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.