missing translation for 'onlineSavingsMsg'
Learn More

(5-Phenylisoxazol-3-yl)methanol, 97%, Thermo Scientific™

Product Code. 10557673
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10557673 250 mg 250mg
10679364 1 g 1g
10317254 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10557673 Supplier Thermo Scientific Maybridge Supplier No. CC30509CB
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1619-37-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD01444104
InChI Key BPAFLGGUEBMWRN-UHFFFAOYSA-N
PubChem CID 1481082
IUPAC Name (5-phenyl-1,2-oxazol-3-yl)methanol
SMILES C1=CC=C(C=C1)C2=CC(=NO2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 5-phenylisoxazol-3-yl methanol, 5-phenyl-3-isoxazolyl methanol, 5-phenyl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol, 5-phenyl, 3-hydroxymethyl-5-phenylisoxazole, 5-phenylisoxazol-3-yl methan-1-ol, 5-phenylisoxazole-3-methanol, 3-isoxazolemethanol,5-phenyl, 5-phenyl-3-hydroxymethylisoxazole, 5-phenyl-isoxazol-3-yl methanol
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material (5-Phenylisoxazol-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.