missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyloxazole, Thermo Scientific™

Product Code. 17750629 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17750629 5 g 5g
17740629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17750629 Supplier Thermo Scientific Acros Supplier No. 264220050
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector PHENYLOXAZOLE
Chemical Name or Material 5-Phenyloxazole
CAS 1006-68-4
Color Brown to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C9 H7 N O
MDL Number 00067074
Quantity 5 g
Molecular Weight (g/mol) 145.16
Physical Form Crystals or Low Melting Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.