missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyltetrazole, 99%

Product Code. 10194061 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194061 5 g Glass bottle 5g
10307481 25 g Glass bottle 25g
10606534 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10194061 Supplier Thermo Scientific Acros Supplier No. 263720050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18039-42-4 | C7H6N4 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18039-42-4
Molecular Formula C7H6N4
Molecular Weight (g/mol) 146.15
MDL Number MFCD00022388
InChI Key MARUHZGHZWCEQU-UHFFFAOYSA-N
Synonym 5-phenyl-1h-tetrazole, 5-phenyltetrazole, 1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, phenyltetrazole, 5-phenyl tetrazole, expandex 5pt, expandex ox 5pt, kempore 50xpt, 5-phenyltetrazole van
PubChem CID 87425
IUPAC Name 5-phenyl-2H-tetrazole
SMILES C1=CC=C(C=C1)C2=NNN=N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 146.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyltetrazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Phenyltetrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.