missing translation for 'onlineSavingsMsg'
Learn More

5α-Pregnane-3β,6α-diol-20-one, MedChemExpress

Product Code. 16497017
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497017 5 mg 5mg
16404308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497017 Supplier Medchem Express Supplier No. HY109564
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone, and it can be produced in starved androgen-responsive prostate cancer cells.

5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone, and it can be produced in starved androgen-responsive prostate cancer cells.

Specifications

Chemical Name or Material 5α-Pregnane-3β,6α-diol-20-one
CAS 21853-11-2
Color White
Purity Grade Notes Research
Molecular Formula C21H34O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (298.96 mM; Need ultrasonic)
SMILES C[C@@]12[C@]3([H])[C@](C[C@H](O)[C@@]1([H])C[C@@H](O)CC2)([H])[C@@]4([H])[C@@](C)([C@@H](C(C)=O)CC4)CC3
Molecular Weight (g/mol) 334.49
Formula Weight 334.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.