missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinecarboxaldehyde, 95%, Thermo Scientific™

Product Code. 15254338 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15254338 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15254338 Supplier Thermo Scientific Acros Supplier No. 454300010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10070-92-5
Melting Point 39.0°C to 43.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C5H4N2O
MDL Number MFCD03426065
Quantity 1 g
Synonym pyrimidine-5-carboxaldehyde, 5-formylpyrimidine, pyrimidine-5-carboxyaldehyde, 5-pyrimidinecarboxaldehyde, 5-pyrimidinecarbaldehyde, 5-pyrimidinaldehyde, 5-formyl pyrimidine, pubchem5298, pyrimidine-5-aldehyde, pyrimidine 5-carboxaldehyde
InChI Key FREJAOSUHFGDBW-UHFFFAOYSA-N
SMILES O=CC1=CN=CN=C1
IUPAC Name pyrimidine-5-carbaldehyde
Molecular Weight (g/mol) 108.10
PubChem CID 2761034
Formula Weight 108.1
Percent Purity 95%
Chemical Name or Material 5-Pyrimidinecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Pyrimidinecarboxaldehyde, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.