missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinylboronic acid, 97%, may contain varying amounts of anhydride, Thermo Scientific Chemicals

Product Code. 15224894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15224894 100 g Glass bottle 100g
15204894 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15224894 Supplier Thermo Scientific Acros Supplier No. 451701000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109299-78-7 | C4H5BN2O2 | 123.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109299-78-7
Molecular Formula C4H5BN2O2
Molecular Weight (g/mol) 123.91
MDL Number MFCD03002366
InChI Key HZFPPBMKGYINDF-UHFFFAOYSA-N
Synonym 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
PubChem CID 2795193
IUPAC Name pyrimidin-5-ylboronic acid
SMILES OB(O)C1=CN=CN=C1

Specifications

Melting Point >300.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 123.91
Percent Purity 97%
Chemical Name or Material 5-Pyrimidinylboronic acid
compliance-icons
Product Identifier
  • 5-Pyrimidinylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.