missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinylboronic acid, 97%, Thermo Scientific™

Product Code. 10624765
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10624765 10 g Amber Glass Bottle 10g
10147712 250 mg Amber Glass Bottle 250mg
10284062 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10624765 Supplier Thermo Scientific Maybridge Supplier No. CC07412EA
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 109299-78-7
Molecular Formula C4H5BN2O2
Molecular Weight (g/mol) 123.91
MDL Number MFCD03002366
InChI Key HZFPPBMKGYINDF-UHFFFAOYSA-N
Synonym 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
PubChem CID 2795193
SMILES OB(O)C1=CN=CN=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name (pyrimidin-5-yl)boronic acid
Formula Weight 123.91
Percent Purity 97%
Chemical Name or Material 5-Pyrimidinylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.