missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrrolidin-1-ylpyridine-2-carboxylic acid, 95%, Thermo Scientific™

Product Code. 11386775
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11386775 250 mg Amber Glass Bottle 250mg
10516704 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386775 Supplier Thermo Scientific Maybridge Supplier No. CC52901CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-19-7
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD09879903
InChI Key ACLFTMQROMOTLA-UHFFFAOYSA-N
Synonym 5-pyrrolidin-1-yl picolinic acid, 5-pyrrolidin-1-yl pyridine-2-carboxylic acid, pubchem19497, 5-pyrrolidinylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid,5-1-pyrrolidinyl, 5-pyrrolidin-1-yl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-1-pyrrolidinyl
PubChem CID 24229530
SMILES OC(=O)C1=NC=C(C=C1)N1CCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 5-pyrrolidin-1-ylpyridine-2-carboxylic acid
Formula Weight 192.22
Percent Purity 95%
Chemical Name or Material 5-Pyrrolidin-1-ylpyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.