missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-SFX (6-(Fluorescein-5-Carboxamido) Hexanoic Acid, Succinimidyl Ester), single isomer

Product Code. 11574826
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11574826 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574826 Supplier Invitrogen™ Supplier No. F6106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-SFX (6-(Fluorescein-5-Carboxamido) Hexanoic Acid, Succinimidyl Ester), single isomer

Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.