missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-193949
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
5965-83-3, 97-05-2
C9H14O6S
250.27
MFCD00007508,MFCD00149540
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
2723734
2-hydroxy-5-sulfobenzoic acid;dihydrate
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
This item is not returnable. View return policy
5965-83-3, 97-05-2
C9H14O6S
250.27
MFCD00007508,MFCD00149540
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
2723734
2-hydroxy-5-sulfobenzoic acid;dihydrate
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 106.0°C to 110.0°C
Color White
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 11, 411
Fieser 01,1118
Merck Index 15, 9094
Solubility Information Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 254.22
Percent Purity 99 to 101%
Grade ACS Reagent
Insoluble Matter 0.02% max
Physical Form Crystalline Powder, Crystals or Granules
Chemical Name or Material 5-Sulfosalicylic acid dihydrate
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'