missing translation for 'onlineSavingsMsg'
Learn More

5-(tert-Butyl)-3-(chloromethyl)-1,2,4-oxadiazole, 95+%, Thermo Scientific™

Product Code. 10602653
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602653 250 mg Amber Glass Bottle 250mg
10099521 1 g Amber Glass Bottle 1g
10173184 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10602653 Supplier Thermo Scientific Maybridge Supplier No. SEW00330CB
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-41-1
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
InChI Key AIVSLDAXUIRICJ-UHFFFAOYSA-N
PubChem CID 2796738
IUPAC Name 5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole
SMILES CC(C)(C)C1=NC(=NO1)CCl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-tert-butyl-3-chloromethyl-1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-1,1-dimethylethyl, 3-chloromethyl-5-t-butyl-1,2,4-oxadiazole, 3-chloromethyl-5-tert-butyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl
Formula Weight 174.63
Percent Purity 95%
Chemical Name or Material 5-(tert-Butyl)-3-(chloromethyl)-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.