missing translation for 'onlineSavingsMsg'
Learn More

5-Thianonane, TRC

Product Code. 30423441
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423441 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423441 Supplier Toronto Research Chemicals Supplier No. TRCO23553350MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutylsulfide
CAS 544-40-1
Molecular Formula C8 H18 S
Quantity 50 mg
Synonym Butyl sulfide (8CI), 1,1'-Thiobis[butane], 5-Thianonane, Butyl monosulfide, Butylthiobutane, Di-n-butyl sulfide, Di-n-butyl thioether, Dibutyl sulfide, Dibutyl thioether, NSC 8460, Thiononane-5, n-Butyl sulfide, n-Dibutyl sulfide
InChI Formula InChI=1S/C8H18S/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3
SMILES CCCCSCCCC
IUPAC Name 1-butylsulfanylbutane
Molecular Weight (g/mol) 146.29
Formula Weight 146.1129
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.