missing translation for 'onlineSavingsMsg'
Learn More

5-Thien-2-yl-1h-pyrazole-3-carboxylic acid, Thermo Scientific™

Product Code. 10472653
Change view
Click to view available options
Quantity:
1 g
5 g
250 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10472653 1 g 1g
10002074 5 g 5g
10127622 250 g 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10472653 Supplier Thermo Scientific Maybridge Supplier No. CC43501DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 182415-24-3
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.208
MDL Number MFCD05170066
InChI Key DUDBNCNNJMOLCC-UHFFFAOYSA-N
Synonym 5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid
PubChem CID 818268
IUPAC Name 5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid
SMILES C1=CSC(=C1)C2=CC(=NN2)C(=O)O

Tekniske data

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 194.21
Percent Purity 97%
Chemical Name or Material 5-Thien-2-yl-1H-pyrazole-3-carboxylic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.